2',3'-Di-O-acetyl-5'-deoxy-5-fluoro-N-(pentyloxycarbonyl)cytidine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: (2R,3R,4R,5R)-2-(5-Fluoro-2-oxo-4-(((pentyloxy)carbonyl)amino)pyrimidin-1(2H)-yl)-5-methyltetrahydrofuran-3,4-diyl diacetate, 5'-Deoxy-5-fluoro-N-[(pentyloxy)carbonyl]cytidine 2',3'-Diacetate

D5787-25G D5787-5G
TCD5787-25GEA 141 USD
TCD5787-25G TCD5787-5G
2',3'-Di-O-acetyl-5'-deoxy-5-fluoro-N-(pentyloxycarbonyl)cytidine ≥98.0% (by HPLC, total nitrogen)
2',3'-Di-O-acetyl-5'-deoxy-5-fluoro-N-(pentyloxycarbonyl)cytidine
Formula: C₁₉H₂₆FN₃O₈
MW: 443.43 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07369303
CAS Number: 162204-20-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 109.0 to 113.0 deg-C
Specific rotation [a]20/D +82.0 to +86.0 deg (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR