Ethyl 2-(3-Formyl-4-hydroxyphenyl)-4-methylthiazole-5-carboxylate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Ethyl 2-(3-formyl-4-hydroxyphenyl)-4-methylthiazole-5-carboxylate

E1394-5G E1394-25G
TCE1394-5GEA 59 USD
TCE1394-5G TCE1394-25G
Ethyl 2-(3-Formyl-4-hydroxyphenyl)-4-methylthiazole-5-carboxylate ≥98.0% (by GC, titration analysis)
Ethyl 2-(3-Formyl-4-hydroxyphenyl)-4-methylthiazole-5-carboxylate
Formula: C₁₄H₁₃NO₄S
MW: 291.32 g/mol
MDL Number: MFCD13194810
CAS Number: 161798-01-2

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 113.0 to 117.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR