N-[2-(Methacryloyloxy)ethyl]-N,N-dimethylbutan-1-aminium bis(trifluoromethanesulfonyl)imide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: Butyl[2-(methacryloyloxy)ethyl](dimethyl)ammonium bis(trifluoromethanesulphonyl)imide

M2987-1G M2987-5G
TCM2987-1GEA 73 USD
TCM2987-1G TCM2987-5G
N-[2-(Methacryloyloxy)ethyl]-N,N-dimethylbutan-1-aminium bis(trifluoromethanesulfonyl)imide ≥98.0% (by HPLC, total nitrogen)
N-[2-(Methacryloyloxy)ethyl]-N,N-dimethylbutan-1-aminium bis(trifluoromethanesulfonyl)imide
Formula: C₁₄H₂₄F₆N₂O₆S₂
MW: 494.46 g/mol
CAS Number: 1616483-28-3

Order Now

Specification Test Results

Appearance Colourless to Light yellow to Light orange clear liquid
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR