(4-Ethynylphenyl)trimethylsilane ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1-Ethynyl-4-(trimethylsilyl)benzene, 4-(Trimethylsilyl)phenylacetylene

E1362-1G E1362-5G
TCE1362-1GEA 130 USD
TCE1362-1G TCE1362-5G
(4-Ethynylphenyl)trimethylsilane ≥98.0% (by GC)
(4-Ethynylphenyl)trimethylsilane
Formula: C₁₁H₁₄Si
Boiling Pt: 75 °C
Storage Temperature: Refrigerator
MDL Number: MFCD26394823
CAS Number: 16116-92-0

Order Now

Specification Test Results

Appearance Colourless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR