Ethyl 2-(3-Cyano-4-isobutoxyphenyl)-4-methyl-5-thiazolecarboxylate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

E1414-25G E1414-5G
TCE1414-25GEA 273 USD
TCE1414-25G TCE1414-5G
Ethyl 2-(3-Cyano-4-isobutoxyphenyl)-4-methyl-5-thiazolecarboxylate ≥98.0% (by HPLC, total nitrogen)
Ethyl 2-(3-Cyano-4-isobutoxyphenyl)-4-methyl-5-thiazolecarboxylate
Formula: C₁₈H₂₀N₂O₃S
MDL Number: MFCD09833701
CAS Number: 160844-75-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 174.0 to 178.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR