1-Ethynyl-4-(trifluoromethoxy)benzene ≥97.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4-(Trifluoromethoxy)phenylacetylene

E1170-1G E1170-5G
TCE1170-1GEA 115 USD
TCE1170-1G TCE1170-5G
1-Ethynyl-4-(trifluoromethoxy)benzene ≥97.0% (by GC)
1-Ethynyl-4-(trifluoromethoxy)benzene
Formula: C₉H₅F₃O
MW: 186.13 g/mol
Flash Pt: 40 °C
MDL Number: MFCD03094333
CAS Number: 160542-02-9
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR