Trifluoromethanesulfonyl bromide ≥95.0% (by GC)

Supplier: TCI America
Danger

Synonyms: Triflyl bromide

T3592-1G T3592-5G
TCT3592-1GEA 62 USD
TCT3592-1G TCT3592-5G
Trifluoromethanesulfonyl bromide ≥95.0% (by GC)
Trifluoromethanesulfonyl bromide
Formula: CBrF₃O₂S
MW: 212.97 g/mol
Storage Temperature: Refrigerator
CAS Number: 15458-53-4
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Appearance Colourless to Brown clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR