(3-Isocyanatopropyl)trimethoxysilane ≥97.0% (by GC)

Supplier: TCI America
Danger

Synonyms: Isocyanic acid 3-(Trimethoxysilyl)propyl ester, 3-(Trimethoxysilyl)propyl Isocyanate

T2044-100G T2044-25G
TCT2044-100GEA 210 USD
TCT2044-100G TCT2044-25G
(3-Isocyanatopropyl)trimethoxysilane ≥97.0% (by GC)
(3-Isocyanatopropyl)trimethoxysilane
Formula: C₇H₁₅NO₄Si
MW: 205.29 g/mol
Boiling Pt: 75 °C
Flash Pt: 108 °C
MDL Number: MFCD00015793
CAS Number: 15396-00-6
EINECS: 239-415-9
UN: 2927
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR