4'-(Di-p-tolylamino)-[1,1'-biphenyl]-4-yl acrylate ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 2-Propenoic acid 4'-[Bis(4-methylphenyl)amino]-[1,1'-biphenyl]-4-yl ester, 4'-[Bis(4-methylphenyl)amino]-[1,1'-biphenyl]-4-yl 2-Propenoate, Acrylic acid 4'-(Di-p-tolylamino)-[1,1'-biphenyl]-4-yl ester

D5507-1G D5507-5G
TCD5507-1GEA 78 USD
TCD5507-1G TCD5507-5G
4'-(Di-p-tolylamino)-[1,1'-biphenyl]-4-yl acrylate ≥97.0% (by HPLC, total nitrogen)
4'-(Di-p-tolylamino)-[1,1'-biphenyl]-4-yl acrylate
Formula: C₂₉H₂₅NO₂
MDL Number: MFCD28975082
CAS Number: 152636-45-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 115.0 to 119.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR