Ethoxytriphenylsilane ≥95.0% (by GC)

Supplier: TCI America
Warning

E1360-1G E1360-5G
TCE1360-1GEA 40 USD
TCE1360-1G TCE1360-5G
Ethoxytriphenylsilane ≥95.0% (by GC)
Ethoxytriphenylsilane
Formula: C₂₀H₂₀OSi
Boiling Pt: 344 °C
MDL Number: MFCD00053752
CAS Number: 1516-80-9
EINECS: 216-170-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 63.0 to 67.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR