2,7-Naphthalenebis(trifluoromethanesulfonate) ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
N1071-1G
N1071-5G
TCN1071-1GEA
64
USD
TCN1071-1G
TCN1071-5G
2,7-Naphthalenebis(trifluoromethanesulfonate) ≥98.0% (by GC)
2,7-Naphthalenebis(trifluoromethanesulfonate)
Formula:
C₁₂H₆F₆O₆S₂ |
MDL Number:
MFCD20260329 CAS Number: 151391-00-3 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 59.0 to 63.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...