(R,R)-2,2'-Isopropylidenebis(4-isopropyl-2-oxazoline) ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (4R,4'R)-2,2'-(Propane-2,2-diyl)bis(4-isopropyl-4,5-dihydrooxazole), (R,R)-2,2-Bis(4-isopropyl-2-oxazolin-2-yl)propane, 2,2-Bis[(4R)-4-isopropyl-2-oxazolin-2-yl]propane

I1089-1G I1089-250MG
TCI1089-1GEA 274 USD
TCI1089-1G TCI1089-250MG
(R,R)-2,2'-Isopropylidenebis(4-isopropyl-2-oxazoline) ≥98.0% (by GC)
(R,R)-2,2'-Isopropylidenebis(4-isopropyl-2-oxazoline)
Formula: C₁₅H₂₆N₂O₂
MW: 266.39 g/mol
MDL Number: MFCD28038687
CAS Number: 150529-94-5

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Optical purity (LC) min. 98.0 ee%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR