1',3'-Dihydro-8-methoxy-1',3',3'-trimethyl-6-nitrospiro[2H-1-benzopyran-2,2'-[2H]indole] ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D5626-1G D5626-5G
TCD5626-1GEA 59 USD
TCD5626-1G TCD5626-5G
1',3'-Dihydro-8-methoxy-1',3',3'-trimethyl-6-nitrospiro[2H-1-benzopyran-2,2'-[2H]indole] ≥98.0% (by titrimetric analysis)
1',3'-Dihydro-8-methoxy-1',3',3'-trimethyl-6-nitrospiro[2H-1-benzopyran-2,2'-[2H]indole]
Formula: C₂₀H₂₀N₂O₄
MDL Number: MFCD00216608
CAS Number: 1498-89-1
EINECS: 216-103-0

Order Now

Specification Test Results

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR