4-(Trifluoromethyl)aniline hydrobromide ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 4-Aminobenzotrifluoride hydrobromide, 4-(Trifluoromethyl)benzenamine hydrobromide, 4-(Trifluoromethyl)anilinium bromide

T3834-1G T3834-5G
TCT3834-1GEA 101 USD
TCT3834-1G TCT3834-5G
4-(Trifluoromethyl)aniline hydrobromide ≥98.0% (by HPLC)
4-(Trifluoromethyl)aniline hydrobromide
Formula: C₇H₆F₃N·HBr
MW: 242.04 g/mol
CAS Number: 148819-81-2

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR