Mono-2-ethylhexyl (2-Ethylhexyl)phosphonate ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: HEHEHP, 2-Ethylhexyl Hydrogen (2-Ethylhexyl)phosphonate, (2-Ethylhexyl)phosphonic acid Mono-2-ethylhexyl ester, PC 88A

M2871-100ML M2871-25ML M2871-5ML
TCM2871-100MLEA 397 USD
TCM2871-100ML TCM2871-25ML TCM2871-5ML
Mono-2-ethylhexyl (2-Ethylhexyl)phosphonate ≥95.0% (by titrimetric analysis)
Mono-2-ethylhexyl (2-Ethylhexyl)phosphonate
Formula: C₁₆H₃₅O₃P
MW: 306.43 g/mol
Flash Pt: 173 °C
MDL Number: MFCD00467188
CAS Number: 14802-03-0
EINECS: 238-865-3
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR