1,3,5-Tris[(1H-imidazol-1-yl)methyl]benzene ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 1,3,5-Tris((1H-imidazol-1-yl)methyl)benzene

T3479-1G T3479-5G
TCT3479-1GEA 114 USD
TCT3479-1G TCT3479-5G
1,3,5-Tris[(1H-imidazol-1-yl)methyl]benzene ≥95.0% (by HPLC, titration analysis)
1,3,5-Tris[(1H-imidazol-1-yl)methyl]benzene
Formula: C₁₈H₁₈N₆
MW: 318.38 g/mol
MDL Number: MFCD32062936
CAS Number: 147951-02-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 183.0 to 187.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR