Benzarone ≥98.0% (by GC)

Supplier: TCI America
Warning

E1289-1G E1289-5G
TCE1289-1GEA 33 USD
TCE1289-1G TCE1289-5G
Benzarone ≥98.0% (by GC)
Benzarone
Formula: C₁₇H₁₄O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00867417
CAS Number: 1477-19-6
EINECS: 216-026-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 124.0 to 128.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR