Formamidine hydrobromide ≥99.0% (by total nitrogen and titration analysis) for perovskite precursor

Supplier: TCI America

F1244-1G F1244-25G F1244-5G
TCF1244-1GEA 33 USD
TCF1244-1G TCF1244-25G TCF1244-5G
Formamidine hydrobromide ≥99.0% (by total nitrogen and titration analysis) for perovskite precursor
Formamidine hydrobromide
Formula: CH₄N₂·HBr
MDL Number: MFCD28369274
CAS Number: 146958-06-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 99.0 %
Purity (Argentmetric Titration) min. 99.0 %
Melting point 133.0 to 137.0 deg-C
Elemental analysis (Carbon) 9.30 to 9.80 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR