N-(9,9-Dimethyl-9H-fluoren-2-yl)-9,9-diphenyl-9H-fluoren-2-amine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

D5588-1G D5588-5G
TCD5588-1GEA 147 USD
TCD5588-1G TCD5588-5G
N-(9,9-Dimethyl-9H-fluoren-2-yl)-9,9-diphenyl-9H-fluoren-2-amine ≥98.0% (by HPLC, total nitrogen)
N-(9,9-Dimethyl-9H-fluoren-2-yl)-9,9-diphenyl-9H-fluoren-2-amine
Formula: C₄₀H₃₁N
MDL Number: MFCD32062889
CAS Number: 1456702-56-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 244.0 to 248.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR