(Butylamine)[2,2'-thiobis(4-tert-octylphenolato)]nickel(II) ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Danger

T3635-25G T3635-5G
TCT3635-25GEA 58 USD
TCT3635-25G TCT3635-5G
(Butylamine)[2,2'-thiobis(4-tert-octylphenolato)]nickel(II) ≥97.0% (by titrimetric analysis)
(Butylamine)[2,2'-thiobis(4-tert-octylphenolato)]nickel(II)
Formula: C₃₂H₅₁NNiO₂S
MDL Number: MFCD03939034
CAS Number: 14516-71-3
EINECS: 238-523-3

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (Chelometric Titration) min. 97.0 %
Lambda max. 246.0 to 254.0 nm (CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR