Eltoprazine dihydrochloride ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
1-(2,3-Dihydro-1,4-benzodioxin-5-yl)piperazine dihydrochloride
E1357-10MG
E1357-50MG
TCE1357-10MGEA
137
USD
TCE1357-10MG
TCE1357-50MG
Eltoprazine dihydrochloride ≥98.0% (by HPLC)
Eltoprazine dihydrochloride
Formula:
C₁₂H₁₆N₂O₂·₂HCl MW: 293.19 g/mol |
CAS Number:
143485-51-2 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...