(4-Hydroxyphenyl)methyl(2-methylbenzyl)sulfonium Hexafluoroantimonate ≥98.0% (by HPLC)

Supplier: TCI America
Warning

H1620-25G H1620-5G
TCH1620-25GEA 429 USD
TCH1620-25G TCH1620-5G
(4-Hydroxyphenyl)methyl(2-methylbenzyl)sulfonium Hexafluoroantimonate ≥98.0% (by HPLC)
(4-Hydroxyphenyl)methyl(2-methylbenzyl)sulfonium Hexafluoroantimonate
Formula: C₁₅H₁₇F₆OSSb
MW: 481.11 g/mol
CAS Number: 141651-31-2
UN: 3077
ADR: 9 III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR