2-([1,1':4',1''-Terphenyl]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
4,4,5,5-Tetramethyl-2-(1,1':4',1''-terphenyl)-3-yl-1,3,2-dioxaborolane, (1,1':4',1''-Terphenyl)-3-ylboronic acid Pinacol ester
T3874-1G
T3874-5G
TCT3874-1GEA
127
USD
TCT3874-1G
TCT3874-5G
2-([1,1':4',1''-Terphenyl]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)
2-([1,1':4',1''-Terphenyl]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula:
C₂₄H₂₅BO₂ MW: 356.27 g/mol |
CAS Number:
1401577-23-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 155.0 to 159.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...