2-Decyl-7-phenyl[1]benzothieno[3,2-b][1]benzothiophene ≥99.5% (by HPLC)
Supplier: TCI America
D5491-100MG
D5491-1G
D5491-250MG
TCD5491-100MGEA
143
USD
TCD5491-100MG
TCD5491-1G
TCD5491-250MG
2-Decyl-7-phenyl[1]benzothieno[3,2-b][1]benzothiophene ≥99.5% (by HPLC)
2-Decyl-7-phenyl[1]benzothieno[3,2-b][1]benzothiophene
Formula:
C₃₀H₃₂S₂ Melting Pt: 225 °C |
CAS Number:
1398395-83-9 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 99.5 area% |
Hole Mobility (mu FET) | min. 10.0 cm2/Vs (ODTS Si/SiO2 substrate) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...