3-[Dimethoxy(methyl)silyl]propyl acrylate ≥95.0% (by GC) stabilized

Supplier: TCI America
Warning

Synonyms: 3-(Dimethoxy(methyl)silyl)propyl acrylate, Acrylic acid 3-[Dimethoxy(methyl)silyl]propyl ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

D5556-25G D5556-5G
TCD5556-25GEA 123 USD
TCD5556-25G TCD5556-5G
3-[Dimethoxy(methyl)silyl]propyl acrylate ≥95.0% (by GC) stabilized
3-[Dimethoxy(methyl)silyl]propyl acrylate
Formula: C₉H₁₈O₄Si
MW: 218.32 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00156431
CAS Number: 13732-00-8

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR