3-(Ethoxydimethylsilyl)propyl methacrylate ≥95.0% (by GC) stabilized
Supplier: TCI America
|
Warning
|
Synonyms:
Methacrylic acid 3-(Ethoxydimethylsilyl)propyl ester, (3-Methacryloyloxy)propyldimethylethoxysilane
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
E1400-1G
E1400-5G
TCE1400-1GEA
88
USD
TCE1400-1G
TCE1400-5G
3-(Ethoxydimethylsilyl)propyl methacrylate ≥95.0% (by GC) stabilized
3-(Ethoxydimethylsilyl)propyl methacrylate
Formula:
C₁₁H₂₂O₃Si MW: 230.38 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00053877 CAS Number: 13731-98-1 |
Specification Test Results
Appearance | Colourless to Almost Colourless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...