1-Fluorocyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

F1254-1G F1254-200MG
TCF1254-1GEA 138 USD
TCF1254-1G TCF1254-200MG
1-Fluorocyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)
1-Fluorocyclopropanecarboxylic acid
Formula: C₄H₅FO₂
MW: 104.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18205954
CAS Number: 137081-41-5
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 72.0 to 76.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR