trans,trans-3,4-Difluoro-4'-(4'-pentylbicyclohexyl-4-yl)biphenyl ≥98.0% (by GC)

Supplier: TCI America

D5651-1G D5651-5G
TCD5651-1GEA 63 USD
TCD5651-1G TCD5651-5G
trans,trans-3,4-Difluoro-4'-(4'-pentylbicyclohexyl-4-yl)biphenyl ≥98.0% (by GC)
trans,trans-3,4-Difluoro-4'-(4'-pentylbicyclohexyl-4-yl)biphenyl
Formula: C₂₉H₃₈F₂
MW: 424.62 g/mol
MDL Number: MFCD11053474
CAS Number: 136609-96-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 95.0 to 263.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR