9,9-Bis(3,4-dicarboxyphenyl)fluorene Dianhydride ≥98.0% (by GC)

Supplier: TCI America

F1192-25G F1192-5G
TCF1192-25GEA 399 USD
TCF1192-25G TCF1192-5G
9,9-Bis(3,4-dicarboxyphenyl)fluorene Dianhydride ≥98.0% (by GC)
9,9-Bis(3,4-dicarboxyphenyl)fluorene Dianhydride
Formula: C₂₉H₁₄O₆
MDL Number: MFCD30342552
CAS Number: 135876-30-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Morpholine Method) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR