Trimethyl(tridecafluorohexyl)silane ≥97.0% (by GC)

Supplier: TCI America
Warning

T3595-1G T3595-5G
TCT3595-1GEA 126 USD
TCT3595-1G TCT3595-5G
Trimethyl(tridecafluorohexyl)silane ≥97.0% (by GC)
Trimethyl(tridecafluorohexyl)silane
Formula: C₉H₉F₁₃Si
MDL Number: MFCD20489406
CAS Number: 135841-49-5
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR