Rifamycin S ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 1,4-Dideoxy-1,4-dihydro-1,4-dioxorifamycin

R0200-1G R0200-5G
TCR0200-1GEA 70 USD
TCR0200-1G TCR0200-5G
Rifamycin S ≥98.0% (by HPLC)
Rifamycin S
Formula: C₃₇H₄₅NO₁₂
MW: 695.76 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06198807
CAS Number: 13553-79-2
EINECS: 236-938-4
Merck Index: 14,08217

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 96.0 %
Specific rotation [a]20/D +451 to +500 deg (C=0.1, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR