Fenpyroximate ≥98.0% (by HPLC)

Supplier: TCI America
Danger

Synonyms: tert-Butyl 4-[[[(E)-[(1,3-Dimethyl-5-phenoxy-1H-pyrazol-4-yl)methylene]amino]oxy]methyl]benzoate, 4-[[[(E)-[(1,3-Dimethyl-5-phenoxy-1H-pyrazol-4-yl)methylene]amino]oxy]methyl]benzoic acid tert-Butyl ester

F1195-100MG F1195-1G
TCF1195-100MGEA 59 USD
TCF1195-100MG TCF1195-1G
Fenpyroximate ≥98.0% (by HPLC)
Fenpyroximate
Formula: C₂₄H₂₇N₃O₄
MW: 421.50 g/mol
MDL Number: MFCD00274596
CAS Number: 134098-61-6
UN: 2811
ADR: 6.1,III
Merck Index: 14,03996

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 9.50 to 10.50 %
Melting point 100.0 to 104.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR