9-Hexyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 2-(9-Hexyl-9H-carbazol-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 9-Hexyl-9H-carbazole-2-boronic acid Pinacol ester

H1647-200MG H1647-1G
TCH1647-200MGEA 72 USD
TCH1647-200MG TCH1647-1G
9-Hexyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥98.0% (by HPLC)
9-Hexyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
Formula: C₂₄H₃₂BNO₂
MW: 377.34 g/mol
CAS Number: 1339953-35-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 83.0 to 87.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR