(S,S)-2,2'-Isopropylidenebis(4-isopropyl-2-oxazoline) ≥98.0% (by GC)

Supplier: TCI America

I1051-1G I1051-250MG
TCI1051-1GEA 186 USD
TCI1051-1G TCI1051-250MG
(S,S)-2,2'-Isopropylidenebis(4-isopropyl-2-oxazoline) ≥98.0% (by GC)
(S,S)-2,2'-Isopropylidenebis(4-isopropyl-2-oxazoline)
Formula: C₁₅H₂₆N₂O₂
MDL Number: MFCD08458310
CAS Number: 131833-92-6

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Optical purity (LC) min. 98.0 ee%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR