2-Hydroxybutyl methacrylate (mixture of isomers) ≥97.0% (by GC) stabilised

Supplier: TCI America

Synonyms: Methacrylic acid 2-Hydroxybutyl ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

H1714-25G H1714-500G
TCH1714-25GEA 36 USD
TCH1714-25G TCH1714-500G
2-Hydroxybutyl methacrylate (mixture of isomers) ≥97.0% (by GC) stabilised
2-Hydroxybutyl methacrylate (mixture of isomers)
Formula: C₈H₁₄O₃
MW: 158.20 g/mol
Boiling Pt: 98 °C
MDL Number: MFCD19707078
CAS Number: 13159-51-8

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (GC) min. 97.0 % (total of isomer)
Acid Value max. 2.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR