5-Oxotetrahydrofuran-3-yl methacrylate ≥97.0% (by GC) stabilized

Supplier: TCI America

Synonyms: beta-Hydroxy-gamma-butyrolactone methacrylate, Methacrylic acid 5-Oxotetrahydrofuran-3-yl ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

O0538-25G O0538-5G
TCO0538-25GEA 302 USD
TCO0538-25G TCO0538-5G
5-Oxotetrahydrofuran-3-yl methacrylate ≥97.0% (by GC) stabilized
5-Oxotetrahydrofuran-3-yl methacrylate
Formula: C₈H₁₀O₄
MW: 170.16 g/mol
Storage Temperature: Refrigerator
CAS Number: 130224-95-2

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR