4,4'-(Ethyne-1,2-diyl)diphthalic anhydride ≥99.0% (by HPLC, titration analysis), purified by sublimation

Supplier: TCI America
Warning

Synonyms: 5,5'-(Ethyne-1,2-diyl)bis(isobenzofuran-1,3-dione)

E1412-1G E1412-5G
TCE1412-1GEA 54 USD
TCE1412-1G TCE1412-5G
4,4'-(Ethyne-1,2-diyl)diphthalic anhydride ≥99.0% (by HPLC, titration analysis), purified by sublimation
4,4'-(Ethyne-1,2-diyl)diphthalic anhydride
Formula: C₁₈H₆O₆
MW: 318.24 g/mol
MDL Number: MFCD28976167
CAS Number: 129808-00-0

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Morpholine Method) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR