4'-Methoxy-N,N-di-p-tolyl-[1,1'-biphenyl]-4-amine ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
4'-Methoxy-N,N-bis(4-methylphenyl)-[1,1'-biphenyl]-4-amine
M3112-1G
M3112-5G
TCM3112-1GEA
32
USD
TCM3112-1G
TCM3112-5G
4'-Methoxy-N,N-di-p-tolyl-[1,1'-biphenyl]-4-amine ≥98.0% (by HPLC, total nitrogen)
4'-Methoxy-N,N-di-p-tolyl-[1,1'-biphenyl]-4-amine
Formula:
C₂₇H₂₅NO |
CAS Number:
129119-41-1 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 141.0 to 145.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...