trans-1-Ethoxy-2,3-difluoro-4-(4-pentylcyclohexyl)benzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Ethoxy-2,3-difluoro-4-(trans-4-pentylcyclohexyl)benzene

E1389-1G E1389-5G
TCE1389-1GEA 41 USD
TCE1389-1G TCE1389-5G
trans-1-Ethoxy-2,3-difluoro-4-(4-pentylcyclohexyl)benzene ≥98.0% (by GC)
trans-1-Ethoxy-2,3-difluoro-4-(4-pentylcyclohexyl)benzene
Formula: C₁₉H₂₈F₂O
MW: 310.43 g/mol
MDL Number: MFCD11053401
CAS Number: 124729-02-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 54.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR