Dicyclohexyl-3,4,3',4'-tetracarboxylic Dianhydride ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

D5723-5G D5723-25G
TCD5723-5GEA 94 USD
TCD5723-5G TCD5723-25G
Dicyclohexyl-3,4,3',4'-tetracarboxylic Dianhydride ≥97.0% (by titrimetric analysis)
Dicyclohexyl-3,4,3',4'-tetracarboxylic Dianhydride
Formula: C₁₆H₁₈O₆
MDL Number: MFCD23134595
CAS Number: 122640-83-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Methanolysis Method) min. 97.0 %
Melting point 206.0 to 210.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR