4-(2-Ethylhexyl)-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 4-(2-Ethylhexyl)-2-thiopheneboronic acid Pinacol ester, 2-[4-(2-Ethylhexyl)-2-thienyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

E1355-1G E1355-5G
TCE1355-1GEA 172 USD
TCE1355-1G TCE1355-5G
4-(2-Ethylhexyl)-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥97.0% (by GC)
4-(2-Ethylhexyl)-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene
Formula: C₁₈H₃₁BO₂S
MW: 322.31 g/mol
Storage Temperature: Refrigerator
CAS Number: 1224430-50-5

Order Now

Specification Test Results

Appearance Colourless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR