4-Glycidyloxy-2,2,6,6-tetramethylpiperidine 1-oxyl ≥95.0% (by GC, titration analysis) free radical

Supplier: TCI America
Warning

Synonyms: 4-Glycidyloxy-TEMPO Free Radical

G0555-1G G0555-5G
TCG0555-1GEA 107 USD
TCG0555-1G TCG0555-5G
4-Glycidyloxy-2,2,6,6-tetramethylpiperidine 1-oxyl ≥95.0% (by GC, titration analysis) free radical
4-Glycidyloxy-2,2,6,6-tetramethylpiperidine 1-oxyl
Formula: C₁₂H₂₂NO₃
MW: 228.31 g/mol
Melting Pt: 38 °C
Storage Temperature: Refrigerator
CAS Number: 122413-85-8

Order Now

Specification Test Results

Appearance Orange to Brown powder to crystal
Purity (GC) min. 95.0 %
Purity (Iodometric Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR