N-Methoxy-N,4-dimethylbenzamide ≥95.0% (by GC)

Supplier: TCI America
Warning

M3098-1G M3098-5G
TCM3098-1GEA 46 USD
TCM3098-1G TCM3098-5G
N-Methoxy-N,4-dimethylbenzamide ≥95.0% (by GC)
N-Methoxy-N,4-dimethylbenzamide
Formula: C₁₀H₁₃NO₂
MDL Number: MFCD02684311
CAS Number: 122334-36-5

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR