3,6-Dibromo-9-(2-naphthalenyl)-9H-carbazole ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

D5546-1G D5546-5G
TCD5546-1GEA 82 USD
TCD5546-1G TCD5546-5G
3,6-Dibromo-9-(2-naphthalenyl)-9H-carbazole ≥98.0% (by HPLC, total nitrogen)
3,6-Dibromo-9-(2-naphthalenyl)-9H-carbazole
Formula: C₂₂H₁₃Br₂N
MDL Number: MFCD29072769
CAS Number: 1221237-83-7

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 197.0 to 201.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR