[Dimethoxy(methyl)silyl]methyl methacrylate ≥90.0% (by GC) stabilised

Supplier: TCI America
Warning

Synonyms: Methacrylic acid [Dimethoxy(methyl)silyl]methyl ester, (Dimethoxy(methyl)silyl)methyl methacrylate

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

D5699-25G D5699-5G
TCD5699-25GEA 251 USD
TCD5699-25G TCD5699-5G
[Dimethoxy(methyl)silyl]methyl methacrylate ≥90.0% (by GC) stabilised
[Dimethoxy(methyl)silyl]methyl methacrylate
Formula: C₈H₁₆O₄Si
Storage Temperature: Refrigerator
MDL Number: MFCD07777082
CAS Number: 121177-93-3

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR