trans,trans-3,4-Difluoro-4'-(4'-propylbicyclohexyl-4-yl)biphenyl ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
3,4-Difluoro-4'-((trans,trans)-4'-propyl-[1,1'-bi(cyclohexan)]-4-yl)-1,1'-biphenyl
D5650-1G
D5650-5G
TCD5650-1GEA
76
USD
TCD5650-1G
TCD5650-5G
trans,trans-3,4-Difluoro-4'-(4'-propylbicyclohexyl-4-yl)biphenyl ≥98.0% (by GC)
trans,trans-3,4-Difluoro-4'-(4'-propylbicyclohexyl-4-yl)biphenyl
Formula:
C₂₇H₃₄F₂ MW: 396.57 g/mol |
MDL Number:
MFCD11053440 CAS Number: 119990-81-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...