1,3,5-Tris(4-formylphenyl)benzene ≥96.0% (by HPLC)

Supplier: TCI America

Synonyms: 5'-(4-Formylphenyl)-[1,1':3',1''-terphenyl]-4,4''-dicarboxaldehyde, 5'-(4-Formylphenyl)-[1,1':3',1''-terphenyl]-4,4''-dicarbaldehyde

F1252-1G F1252-200MG
TCF1252-1GEA 331 USD
TCF1252-1G TCF1252-200MG
1,3,5-Tris(4-formylphenyl)benzene ≥96.0% (by HPLC)
1,3,5-Tris(4-formylphenyl)benzene
Formula: C₂₇H₁₈O₃
MW: 390.44 g/mol
MDL Number: MFCD27937712
CAS Number: 118688-53-2

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 96.0 area%
Melting point 248.0 to 252.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR