1-Propoxy-4-[[4-(trans-4-propylcyclohexyl)phenyl]ethynyl]benzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-[(4-Propoxyphenyl)ethynyl]-4-(trans-4-propylcyclohexyl)benzene

P2440-1G P2440-5G
TCP2440-1GEA 70 USD
TCP2440-1G TCP2440-5G
1-Propoxy-4-[[4-(trans-4-propylcyclohexyl)phenyl]ethynyl]benzene ≥98.0% (by GC)
1-Propoxy-4-[[4-(trans-4-propylcyclohexyl)phenyl]ethynyl]benzene
Formula: C₂₆H₃₂O
MDL Number: MFCD11053434
CAS Number: 116903-49-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 96 to 233 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR