4-Fluoro-N-methoxy-N-methylbenzamide ≥97.0% (by GC)

Supplier: TCI America

F1194-1G F1194-5G
TCF1194-1GEA 44 USD
TCF1194-1G TCF1194-5G
4-Fluoro-N-methoxy-N-methylbenzamide ≥97.0% (by GC)
4-Fluoro-N-methoxy-N-methylbenzamide
Formula: C₉H₁₀FNO₂
Boiling Pt: 120 °C
MDL Number: MFCD02179265
CAS Number: 116332-54-8

Order Now

Specification Test Results

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR