Sodium Pyridine-4-sulfinate ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

O0539-1G O0539-250MG
TCO0539-1GEA 121 USD
TCO0539-1G TCO0539-250MG
Sodium Pyridine-4-sulfinate ≥95.0% (by HPLC, titration analysis)
Sodium Pyridine-4-sulfinate
Formula: C₅H₄NNaO₂S
MDL Number: MFCD30502924
CAS Number: 116008-37-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR